missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroazetidine hydrochloride, 95%

Product Code. 15502313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15502313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15502313 Supplier Thermo Scientific Acros Supplier No. 449990010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 617718-46-4 | C3H6FN·HCl | 111.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 617718-46-4
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass Bottle
Molecular Formula C3H6FN·HCl
Quantity 1 g
Synonym 3-fluoroazetidine hydrochloride, 3-fluoroazetidine hcl, 3-fluoroazetidinehydrochloride, azetidine, 3-fluoro-, hydrochloride, 3-fluoroazetane hydrochloride, acmc-20dlgf, c3h6fn.hcl, ksc941o8n, 3-fluoro-azetidine hydrochloride, 3-fluoroazetidine hcl salt
InChI Key PXFUWRWCKSLCLS-UHFFFAOYSA-N
SMILES C1C(CN1)F.Cl
IUPAC Name 3-fluoroazetidine;hydrochloride
Molecular Weight (g/mol) 111.55
PubChem CID 10125054
Formula Weight 111.55
Percent Purity 95%
Chemical Name or Material 3-Fluoroazetidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluoroazetidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.