missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophthalic anhydride, 98%, ACROS Organics™

Product Code. 10689834 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10689834 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10689834 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 293700010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 652-39-1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 159°C to 163°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H3FO3
MDL Number MFCD00039696
Quantity 1 g
Synonym 3-fluorophthalic anhydride, 4-fluoroisobenzofuran-1,3-dione, 3-fluorophthalicanhydride, 4-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 4-fluoro, 5-fluoro-isobenzofurandione, 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluor-2-benzofuran-1,3-dion, pubchem1949, fluorophthalic anhydride
InChI Key WWJAZKZLSDRAIV-UHFFFAOYSA-N
SMILES FC1=CC=CC2=C1C(=O)OC2=O
IUPAC Name 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 166.11
PubChem CID 69551
Formula Weight 166.11
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorophthalic anhydride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorophthalic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.