missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropiperidine hydrochloride, 97%

Product Code. 11319735
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319735 250 mg 250mg
11309735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319735 Supplier Thermo Scientific Alfa Aesar Supplier No. L19776.MD
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 737000-77-0 | C5H11ClFN | 139.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737000-77-0
Molecular Formula C5H11ClFN
Molecular Weight (g/mol) 139.598
MDL Number MFCD03452799
InChI Key RDJUBLSLAULIAT-UHFFFAOYSA-N
Synonym 3-fluoropiperidine hydrochloride, 3-fluoropiperidine hcl, piperidine, 3-fluoro-, hydrochloride, pubchem24370, acmc-20an39, 3-fluoropiperidinehydrochloride, 3-fluoro-piperidine hydrochloride, 3-fluoranylpiperidine hydrochloride, 3-fluoro-piperidine, hydrochloride
PubChem CID 2759156
IUPAC Name 3-fluoropiperidine;hydrochloride
SMILES C1CC(CNC1)F.Cl

Specifications

Melting Point ∼227°C
Quantity 250 mg
Beilstein 6145604
Sensitivity Hygroscopic
Formula Weight 139.6
Percent Purity 97%
Chemical Name or Material 3-Fluoropiperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.