missing translation for 'onlineSavingsMsg'
Learn More

3-Furancarboximidamide hydrochloride, Technical Grade, Thermo Scientific™

Product Code. 10110792
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
10110792 250 mg 250mg
10120792 1 g 1g
10623645 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 10110792

Brand: Thermo Scientific Maybridge MO00101CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 54649-21-7
Molecular Formula C5H7ClN2O
Molecular Weight (g/mol) 146.574
InChI Key RJLCJJIXGGSRPZ-UHFFFAOYSA-N
Synonym furan-3-carboximidamide hydrochloride, 3-furancarboximidamide hydrochloride, furan-3-carboxamidine hcl, furan-3-carboxamidine, chloride, 3-furancarboximidamide,hydrochloride 1:1
PubChem CID 2779666
IUPAC Name furan-3-carboximidamide;hydrochloride
SMILES C1=COC=C1C(=N)N.Cl

Specifications

Quantity 250 mg
Formula Weight 146.58
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Furancarboximidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.