missing translation for 'onlineSavingsMsg'
Learn More

3-Glycidoxypropyltrimethoxysilane, 97%

Product Code. 10697012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697012 500 g Glass bottle 500g
10791101 5 g Glass bottle 5g
10402221 100 g Glass bottle 100g
11985821 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10697012 Supplier Thermo Scientific Acros Supplier No. 216545000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2530-83-8 | C9H20O5Si | 236.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-83-8
Molecular Formula C9H20O5Si
Molecular Weight (g/mol) 236.34
InChI Key BPSIOYPQMFLKFR-UHFFFAOYSA-N
Synonym 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
PubChem CID 17317
IUPAC Name trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](CCCOCC1CO1)(OC)OC

Specifications

Density 1.0700g/mL
Boiling Point 120.0°C (2.0 mmHg)
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4280 to 1.4300
Quantity 500 g
Beilstein 17, V,3, 45
Specific Gravity 1.07
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, benzene and ether
Viscosity 4 mPa.s (20°C)
Formula Weight 236.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Glycidoxypropyltrimethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • 3-Glycidoxypropyltrimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.