missing translation for 'onlineSavingsMsg'
Learn More

3-Guanidinopropionic acid, 97%

Product Code. 10264644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10264644 5 g 5g
10051374 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10264644 Supplier Thermo Scientific Acros Supplier No. 426660050
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 353-09-3 | C4H9N3O2 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-09-3
Molecular Formula C4H9N3O2
Molecular Weight (g/mol) 131.13
MDL Number MFCD00045939
InChI Key KMXXSJLYVJEBHI-UHFFFAOYSA-N
Synonym 3-guanidinopropionic acid, 3-guanidinopropanoic acid, beta-guanidinopropionic acid, guanidinopropionic acid, 3-guanidinopropanoate, 3-carbamimidamidopropanoic acid, beta-gpa, unii-ul1984yrka, beta-guanadinopropionate, n-aminoiminomethyl-beta-alanine
PubChem CID 67701
ChEBI CHEBI:15968
IUPAC Name 3-(diaminomethylideneamino)propanoic acid
SMILES C(CN=C(N)N)C(=O)O

Specifications

Melting Point 222.0°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)2CO2H
Quantity 5 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material 3-Guanidinopropionic acid
compliance-icons
Product Identifier
  • 3-Guanidinopropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.