missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanone, 98%

Product Code. 11418816
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418816 1000 mL 1000mL
11428816 50 mL 50mL
11438816 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418816 Supplier Thermo Scientific Alfa Aesar Supplier No. A16977.AU
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-35-4 | C7H14O | 114.19 g/mol

Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

Solubility
Soluble in water at 20°C 4.3g/L.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-35-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009483
InChI Key NGAZZOYFWWSOGK-UHFFFAOYSA-N
Synonym 3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
PubChem CID 7802
ChEBI CHEBI:50139
IUPAC Name heptan-3-one
SMILES CCCCC(=O)CC

Specifications

Melting Point -37°C
Density 0.818
Boiling Point 147°C to 149°C
Flash Point 41°C (105°F)
Odor Pungent
Refractive Index 1.409
Quantity 1000 mL
UN Number UN1224
Beilstein 506161
Solubility Information Soluble in water at 20°C 4.3g/L.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 3-Heptanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.