missing translation for 'onlineSavingsMsg'
Learn More

3-Hepten-2-one, 95%

Product Code. 11449386
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449386 10 g 10g
11459386 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449386 Supplier Thermo Scientific Alfa Aesar Supplier No. A19086.09
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1119-44-4 | C7H12O | 112.17 g/mol

It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

Solubility
Soluble in alcohol and slightly soluble in water(3087 mg/L @ 25°C (est.))

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-44-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00015564
InChI Key JHHZQADGLDKIPM-AATRIKPKSA-N
Synonym 3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one
PubChem CID 5364578
SMILES CCC\C=C\C(C)=O

Specifications

Density 0.86
Boiling Point 156°C
Flash Point 49°C (120°F)
Refractive Index 1.445
Quantity 10 g
UN Number UN1224
Beilstein 1739239
Solubility Information Soluble in alcohol and slightly soluble in water(3087mg/L @ 25°C (est.))
IUPAC Name (E)-hept-3-en-2-one
Formula Weight 112.16
Percent Purity 97%
Chemical Name or Material 3-Hepten-2-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.