missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-2-one, 97%, Thermo Scientific™

Product Code. 10423623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10423623 1 g Glass Bottle 1g
10006232 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423623 Supplier Thermo Scientific Acros Supplier No. 379700010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1679-36-3
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00041627
InChI Key LTAPKZGQTMVYMX-UHFFFAOYSA-N
Synonym 3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
PubChem CID 137151
IUPAC Name hex-3-yn-2-one
SMILES CCC#CC(=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.8780g/mL
Boiling Point 68.0°C (50.0 mmHg)
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3CH2C≡CCOCH3
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.878
Formula Weight 96.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Hexyn-2-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.