missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyne, 99%

Product Code. 11327795
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327795 25 g 25g
11317795 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327795 Supplier Thermo Scientific Alfa Aesar Supplier No. L02682.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-49-4 | C6H10 | 82.15 g/mol

3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hexyne is used in the synthesis of fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety by reacting with ruthenacycles. It was also used in the preparation of [4+2] cycloaddition product by reacting with borole.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-49-4
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00009381
InChI Key DQQNMIPXXNPGCV-UHFFFAOYSA-N
Synonym 3-hexyne, diethylacetylene, unii-9gtq990q4k, acmc-209rhw, c2h5c.$.cc2h5
PubChem CID 13568
IUPAC Name hex-3-yne
SMILES CCC#CCC

Specifications

Density 0.721
Boiling Point 80°C to 82°C
Flash Point −10°C (14°F)
Odor Characteristic
Linear Formula CH3CH2C≡CCH2CH3
Refractive Index 1.411
Quantity 25 g
UN Number UN3295
Beilstein 1731158
Solubility Information Not miscible in water.
Formula Weight 82.15
Percent Purity 99%
Chemical Name or Material 3-Hexyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.