missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-methyl-5,6-indolinedione, 98%

Product Code. 10784185 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10784185 250 mg Glass bottle 250mg
10654945 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10784185

Brand: Thermo Scientific Acros 426212500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54-06-8 | C9H9NO3 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-06-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.17
MDL Number MFCD00069732
InChI Key RPHLQSHHTJORHI-UHFFFAOYSA-N
Synonym adrenochrome, adraxone, usaf uctl-7, 3-hydroxy-1-methyl-5,6-indolinedione, 1-adrenochrome, 3-hydroxy-1-methylindoline-5,6-dione, 2,3-dihydro-3-hydroxy-1-methyl-1h-indole-5,6-dione, 5,6-indolinedione, 3-hydroxy-1-methyl, adrenochrome, l, adrenochrom
PubChem CID 5898
IUPAC Name 3-hydroxy-1-methyl-2,3-dihydroindole-5,6-dione
SMILES CN1CC(C2=CC(=O)C(=O)C=C21)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 161
Formula Weight 179.17
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-1-methyl-5, 6-indolinedione
compliance-icons
Product Identifier
  • 3-Hydroxy-1-methyl-5,6-indolinedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.