missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride, 85%, technical, Thermo Scientific™

Product Code. 15289616 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15289616 25 g 25g
15279616 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15289616 Supplier Thermo Scientific Acros Supplier No. 121260250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14173-30-9
Molecular Formula C6H7NO2·HCl
Molecular Weight (g/mol) 161.59
InChI Key SKVRYQUMKJQZFN-UHFFFAOYSA-N
Synonym 3-hydroxy-2-pyridinemethanol hydrochloride, 2-hydroxymethyl pyridin-3-ol hydrochloride, 3-hydroxy-2-hydroxymethyl pyridine hydrochloride, 2-pyridinemethanol, 3-hydroxy-, hydrochloride, 2-hydroxymethyl pyridin-3-ol hcl, 3-hydroxy-2-hydroxymethylpyridine hydrochloride, pubchem4075, 3-hydroxy-2-hydroxymethyl pyridine hcl, acmc-209xde, 3-hydroxy-2-pyridinemethanol hcl
PubChem CID 3084185
IUPAC Name 2-(hydroxymethyl)pyridin-3-ol;hydrochloride
SMILES C1=CC(=C(N=C1)CO)O.Cl

Specifications

Melting Point 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 84% min. (HPLC)
Linear Formula HO(C5H3N)CH2OH
Quantity 25 g
Formula Weight 161.59
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride
compliance-icons
Product Identifier
  • 3-Hydroxy-2-(hydroxymethyl)pyridine hydrochloride, 85%, Technical
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.