missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-nitropyridine, 98%

Product Code. 11369479
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369479 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11369479 Supplier Thermo Scientific Alfa Aesar Supplier No. A14887.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15128-82-2 | C5H4N2O3 | 140.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 69°C to 73°C
CAS 15128-82-2
Quantity 100 g
Molecular Formula C5H4N2O3
Synonym 2-Nitro-3-pyridinol;
InChI Key QBPDSKPWYWIHGA-UHFFFAOYSA-N
SMILES OC1=CC=CN=C1[N+]([O-])=O
IUPAC Name 2-nitropyridin-3-ol
Molecular Weight (g/mol) 140.10
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.