missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-pyrrolidinone, 95%, Thermo Scientific Chemicals

Product Code. 15404358
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404358 1 g 1g
15414358 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404358 Supplier Thermo Scientific Alfa Aesar Supplier No. H57961.03
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

3-Hydroxy-2-pyrrolidinone is used as a potent inhibitor of CDK2/cyclin A. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-2-pyrrolidinone is used as a potent inhibitor of CDK2/cyclin A. It is also used as pharmaceutical intermediates.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Store under dry inert gas. Incompatible with heat, oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15166-68-4
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD09751256
InChI Key FRKGSNOMLIYPSH-UHFFFAOYSA-N
Synonym 3-hydroxy-2-pyrrolidinone, 2-pyrrolidinone, 3-hydroxy, 3-hydroxy-pyrrolidin-2-one, a-hydroxy-?-butyrolactam, acmc-209pe0, 3-oxidanylpyrrolidin-2-one, 3-hydroxy-2-oxopyrrolidine, 3-hydroxypyrrolidine-2-one
PubChem CID 10240772
IUPAC Name 3-hydroxypyrrolidin-2-one
SMILES C1CNC(=O)C1O

Specifications

Melting Point 102°C to 107°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 101.11
Percent Purity 95%
Chemical Name or Material 3-Hydroxy-2-pyrrolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.