missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methyl-2-butanone, 92%

Product Code. 10169072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10169072 5 g Glass bottle 5g
10428080 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10169072 Supplier Thermo Scientific Acros Supplier No. 121440050
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 115-22-0 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-22-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00004460
InChI Key BNDRWEVUODOUDW-UHFFFAOYSA-N
Synonym 3-hydroxy-3-methyl-2-butanone, 3-methylacetoin, methylacetoin, 2-butanone, 3-hydroxy-3-methyl, dimethylacetylcarbinol, 2-hydroxy-2-methyl-3-butanone, 2-methyl-2-hydroxybutan-3-one, 1-hydroxy-1-methylethyl methyl ketone, 3-hydroxy-3-methyl-butan-2-one, 3-hydroxy-3-ethylbutanone
PubChem CID 8261
IUPAC Name 3-hydroxy-3-methylbutan-2-one
SMILES CC(=O)C(C)(C)O

Specifications

CAS Min % 90.0
CAS Max % 100.0
Color Colorless to Yellow or Red
Density 0.9700g/mL
Boiling Point 140°C to 141°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C(OH)COCH3
Refractive Index 1.4145 to 1.4165
Quantity 5 g
Beilstein 01, 832
Specific Gravity 0.97
Formula Weight 102.13
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone, 92%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-3-methyl-2-butanone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.