missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxyphenylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11906111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11906111 1 g 1g
11926111 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11906111 Supplier Thermo Scientific Acros Supplier No. 445060010
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 269410-23-3
Molecular Formula C13H19BO4
Molecular Weight (g/mol) 250.10
MDL Number MFCD09027077
InChI Key WINSAMQIAMEZIK-UHFFFAOYSA-N
Synonym 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxy-4-methoxyphenylboronic acid, pinacol ester, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxy-4-methoxyphenylboronic acid pinacol ester, phenol, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb953, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenol
PubChem CID 17750251
IUPAC Name 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES COC1=CC=C(C=C1O)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 101.0°C to 102.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 250.1
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-4-methoxyphenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Hydroxy-4-methoxyphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.