missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methyl-2(5H)-thiophenone, TRC

Product Code. 30137703
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137703 1 g 1g
30009221 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137703 Supplier Toronto Research Chemicals Supplier No. TRCH8255451G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34876-35-2
Molecular Formula C5 H6 O2 S
Molecular Weight (g/mol) 130.165
Synonym 3-Hydroxy-4-methyl-2(5H)-thiophenone, Cefadroxil Monohydrate Imp. G (EP), Cefalexin Imp. D (EP), Cefradine Imp. F (EP)
IUPAC Name 4-hydroxy-3-methyl-2H-thiophen-5-one
SMILES CC1=C(O)C(=O)SC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H6O2S/c1-3-2-8-5(7)4(3)6/h6H,2H2,1H3
Formula Weight 130.009
Chemical Name or Material 3-Hydroxy-4-methyl-2(5H)-thiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.