missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-5-methylpyridine, 97%

Product Code. 15425407
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15425407

Brand: Thermo Scientific Alfa Aesar H32658.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42732-49-0 | C6H7NO | 109.128 g/mol

3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42732-49-0
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00661297
InChI Key RYJNCIGFPWGVPA-UHFFFAOYSA-N
Synonym 3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg
PubChem CID 224753
IUPAC Name 5-methylpyridin-3-ol
SMILES CC1=CC(=CN=C1)O

Specifications

Melting Point 136°C to 140°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-5-methylpyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.