missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Anagrelide-13C3 (~80%), TRC

Product Code. 30395994
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395994 0.5 mg 0.5mg
30406182 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395994 Supplier Toronto Research Chemicals Supplier No. TRCH7983030.5MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy Anagrelide-13C3
CAS 1219393-12-0
Molecular Formula C713C3H7Cl2N3O2
Quantity 0.5 mg
Synonym 6,7-Dichloro-1,5-dihydro-3-hydroxyimidazo[2,1-b]quinazolin-2(3H)-one-13C3, BCH 24426-13C3
InChI Formula InChI=1S/C10H7Cl2N3O2/c11-5-1-2-6-4(7(5)12)3-15-9(17)8(16)14-10(15)13-6/h1-2,9,17H,3H2,(H,13,14,16)/i8+1,9+1,10+1
SMILES ClC1=C(Cl)C=CC2=C1CN([13CH](O)[13C](N3)=O)[13C]3=N2
Molecular Weight (g/mol) 275.07
Formula Weight 274.0016
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.