missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Desloratadine, TRC

Product Code. 30406914
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406914 50 mg
30392867 1 mg
30399509 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406914 Supplier Toronto Research Chemicals Supplier No. TRCH93675050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119410-08-1
Molecular Formula C19 H19 Cl N2 O
Molecular Weight (g/mol) 326.82
Synonym 5H-Benzo[5,6]cyclohepta[1,2-b]pyridin-3-ol, 8-chloro-6,11-dihydro-11-(4-piperidinylidene)-, 8-Chloro-6,11-dihydro-11-(4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-3-ol, 3-Hydroxydesloratadine, Sch 45581
IUPAC Name 8-chloro-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-3-ol
SMILES Oc1cnc2C(=C3CCNCC3)c4ccc(Cl)cc4CCc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H19ClN2O/c20-15-3-4-17-13(9-15)1-2-14-10-16(23)11-22-19(14)18(17)12-5-7-21-8-6-12/h3-4,9-11,21,23H,1-2,5-8H2
Formula Weight 326.1186
Percent Purity >95%
Chemical Name or Material 3-Hydroxy Desloratadine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.