missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyadamantane-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10161292
Change view
Click to view available options
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10161292 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10161292 Supplier Thermo Scientific Maybridge Supplier No. BTB12757EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 42711-75-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H15O3
MDL Number MFCD00193929
Quantity 10 g
Synonym 3-hydroxy-1-adamantanecarboxylic acid, 3-hydroxy-adamantane-1-carboxylic acid, 3-hydroxyadamantanecarboxylic acid, 1-carboxy-3-adamantanol, 3-hydroxy-1-adamantane carboxylic acid, 3-hydroxytricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3-carboxy-1-hydroxyadamantane, 3-hydroxy-tricyclo, acmc-1aqmu, 3-carboxy-1-adamantanol
InChI Key CJJMAWPEZKYJAP-JZVMUCMXSA-M
SMILES OC12C[C@@H]3C[C@H](C1)CC(C3)(C2)C([O-])=O
IUPAC Name 3-hydroxyadamantane-1-carboxylic acid
Molecular Weight (g/mol) 195.24
PubChem CID 2736518
Formula Weight 196.24
Percent Purity 97%
Chemical Name or Material 3-Hydroxyadamantane-1-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.