missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybutyric acid, 95%

Product Code. 10192530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10192530 5 g Glass bottle 5g
10245110 10 g Glass bottle 10g
10239743 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10192530 Supplier Thermo Scientific Acros Supplier No. 121050050
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 300-85-6 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-85-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004546
InChI Key WHBMMWSBFZVSSR-UHFFFAOYSA-N
Synonym 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy
PubChem CID 441
ChEBI CHEBI:20067
IUPAC Name 3-hydroxybutanoic acid
SMILES CC(CC(=O)O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 1.1260g/mL
Boiling Point 118.0°C to 120.0°C (2.0 mmHg)
Flash Point 112°C
Assay Percent Range 95&percent;
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2CO2H
Refractive Index 1.4420 to 1.4440
Quantity 5 g
Beilstein 03, 307
Fieser 14,178
Merck Index 15, 4855
Specific Gravity 1.126
Solubility Information Solubility in water: >10% in water. Other solubilities: soluble in alcohol,ether
Formula Weight 104.11
Percent Purity 95%
Grade Technical
Chemical Name or Material 3-Hydroxybutyric acid
Show More Show Less
compliance-icons
Identifiant produit
  • 3-Hydroxybutyric acid
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.