missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybutyric acid, tech.

Product Code. 11369528
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369528 25 g 25g
11359528 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369528 Supplier Thermo Scientific Alfa Aesar Supplier No. 043354.14
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 300-85-6 | C4H8O3 | 104.105 g/mol

Industrially, 3-hydroxybutanoic acid is used for the manufacture of biodegradable plastics. It is also acts as a solvent in organoleptics, cosmetics. It is also used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by Escherichia coli.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-85-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00004546
InChI Key WHBMMWSBFZVSSR-UHFFFAOYSA-N
Synonym 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy
PubChem CID 441
ChEBI CHEBI:20067
IUPAC Name 3-hydroxybutanoic acid
SMILES CC(CC(=O)O)O

Specifications

Color Colorless
Density 1.13 g/mL
Boiling Point 130°C (12 mmHg)
Flash Point >112°C (234°F)
Refractive Index 1.4445
Quantity 25 g
Sensitivity Hygroscopic
Merck Index 14,4816
Solubility Information Soluble in water, alcohol, ether
Formula Weight 104.1
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Hydroxybutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.