missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyhexanoic Acid, TRC

Product Code. 30394201
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30394201 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394201 Supplier Toronto Research Chemicals Supplier No. TRCH82532025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxyhexanoic Acid
CAS 10191-24-9
Purity Grade Notes GC
Molecular Formula C6 H12 O3
Quantity 25 mg
Synonym (+/-)-3-Hydroxyhexanoic Acid, 3-Hydroxycaproic Acid, 3-Hydroxyhexanoic Acid, β-Hydroxy-n-caproic Acid, β-Hydroxycaproic Acid, β-Hydroxyhexanoic Acid
InChI Formula InChI=1S/C6H12O3/c1-2-3-5(7)4-6(8)9/h5,7H,2-4H2,1H3,(H,8,9)
SMILES CCCC(O)CC(=O)O
IUPAC Name 3-hydroxyhexanoic acid
Molecular Weight (g/mol) 132.158
Formula Weight 132.0786
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.