missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphthalic Anhydride, TRC

Product Code. 30405018
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405018 2.5 g 2.5g
30408612 5 g -
30405904 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405018 Supplier Toronto Research Chemicals Supplier No. TRCH9512702.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37418-88-5
Molecular Formula C8H4O4
Molecular Weight (g/mol) 164.11
Synonym 3-Hydroxyphthalic Acid Anhydride, 4-Hydroxy-1,3-isobenzofurandione, 4-Hydroxy-1,3-dihydro-2-benzofuran-1,3-dione, 4-Hydroxy-2-benzofuran-1,3-dione, 4-Hydroxyisobenzofuran-1,3-dione, NSC 80858
IUPAC Name 4-hydroxy-2-benzofuran-1,3-dione
SMILES Oc1cccc2C(=O)OC(=O)c12

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H4O4/c9-5-3-1-2-4-6(5)8(11)12-7(4)10/h1-3,9H
Formula Weight 164.011
Chemical Name or Material 3-Hydroxyphthalic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.