missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypiperidine, 98+%

Product Code. 11344564
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344564 50 g 50g
11324564 2 g 2g
11334564 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344564 Supplier Thermo Scientific Alfa Aesar Supplier No. L01333.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6859-99-0 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6859-99-0
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014591
InChI Key BIWOSRSKDCZIFM-UHFFFAOYSA-N
Synonym 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine
PubChem CID 23293
IUPAC Name piperidin-3-ol
SMILES C1CC(CNC1)O

Specifications

Melting Point 56°C to 60°C
pH 11.5 to 11.7
Boiling Point 67°C to 69°C (2 mmHg)
Flash Point >100°C (212°F)
Odor Amine-like
Quantity 50 g
UN Number UN3263
Beilstein 102696
Sensitivity Air sensitive; Hygroscopic
Formula Weight 101.15
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxypiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.