missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypropionitrile, 97%

Product Code. 10553311 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10553311

Brand: Thermo Scientific Acros 149390500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-78-4 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-78-4
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00002826
InChI Key WSGYTJNNHPZFKR-UHFFFAOYSA-N
Synonym 3-hydroxypropionitrile, hydracrylonitrile, 2-cyanoethanol, ethylene cyanohydrin, propanenitrile, 3-hydroxy, beta-cyanoethanol, glycol cyanohydrin, methanolacetonitrile, hydroxypropanenitrile, 2-hydroxycyanoethane
PubChem CID 8011
IUPAC Name 3-hydroxypropanenitrile
SMILES C(CO)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -46°C
Color Yellow
Density 1.0400g/mL
Boiling Point 228°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH2CN
Refractive Index 1.4246 to 1.4266
Quantity 50 g
Beilstein 03, 298
Merck Index 15, 3848
Specific Gravity 1.04
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetone,methyl ethyl ketone and,ethyl alcohol,slightly soluble in ether,insoluble in benzene,carbon disulfide and ccl4
Viscosity <10 mPa.s (20°C)
Formula Weight 71.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Hydroxypropionitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.