missing translation for 'onlineSavingsMsg'
Learn More

3’-Hydroxytyrosol 3’-Glucuronide, TRC

Product Code. 30012138
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012138 1 mg 1mg
30008803 2.5 mg 2.5mg
30006521 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012138 Supplier Toronto Research Chemicals Supplier No. TRCH9770301MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 425408-50-0
Molecular Formula C14H18O9
Molecular Weight (g/mol) 330.29
Synonym 2-Hydroxy-5-(2-hydroxyethyl)phenyl β-D-Glucopyranosiduronic Acid
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-hydroxy-5-(2-hydroxyethyl)phenoxy)tetrahydro-2H-pyran-2-carboxylic acid
SMILES O([C@@H]1O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]1O)C2=CC(CCO)=CC=C2O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H18O9/c15-4-3-6-1-2-7(16)8(5-6)22-14-11(19)9(17)10(18)12(23-14)13(20)21/h1-2,5,9-12,14-19H,3-4H2,(H,20,21)/t9-,10-,11+,12-,14+/m0/s1
Formula Weight 330.1
Chemical Name or Material 3'-Hydroxytyrosol 3'-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.