missing translation for 'onlineSavingsMsg'
Learn More

3-Indazolinone, 97%

Product Code. 11367615
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367615 5 g 5g
11377615 25 g 25g
11459820 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367615 Supplier Thermo Scientific Alfa Aesar Supplier No. L05860.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7364-25-2 | C7H6N2O | 134.138 g/mol

3-Indazolinone is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Indazolinone is used as catalytic agent, petrochemical additive.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7364-25-2
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00005685
InChI Key SWEICGMKXPNXNU-UHFFFAOYSA-N
Synonym 1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol
PubChem CID 81829
IUPAC Name 1,2-dihydroindazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NN2

Specifications

Melting Point 248°C to 252°C
Quantity 5 g
Beilstein 3733
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 3-Indazolinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.