missing translation for 'onlineSavingsMsg'
Learn More

3-Indazolinone, 97%

Product Code. p-7044322
Click to view available options
Quantity:
5 g
25 g
100 g
Packungsgröße:
100g
25g
5g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11367615

Marke: Thermo Scientific Alfa Aesar L05860.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 7364-25-2 | C7H6N2O | 134.138 g/mol

3-Indazolinone is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Indazolinone is used as catalytic agent, petrochemical additive.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7364-25-2
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00005685
InChI Key SWEICGMKXPNXNU-UHFFFAOYSA-N
Synonym 1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol
PubChem CID 81829
IUPAC Name 1,2-dihydroindazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NN2

Spezifikation

Melting Point 248°C to 252°C
Quantity 5 g
Beilstein 3733
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 3-Indazolinone

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.