missing translation for 'onlineSavingsMsg'
Learn More

3-Indolepropionic Acid, TRC

Product Code. 30394320
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394320 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394320 Supplier Toronto Research Chemicals Supplier No. TRCI62717010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Indolyl)-propionic acid
CAS 830-96-6
Molecular Formula C11 H11 N O2
Quantity 10 g
Synonym Indole-3-propionic acid (8CI), 1H-Indole-3-propionic acid, 3-(1H-Indol-3-yl)propanoic acid, 3-(1H-Indol-3-yl)propionic acid, 3-(2-Carboxyethyl)-1H-indole, 3-(3-Indolyl)propanoic acid, 3-(3-Indolyl)propionic acid, 3-(Indole-3-yl)propanoic acid, IPA, IPA (auxin), NSC 3252, NSC 47831, β-(3-Indolyl)propionic acid, β-Indole-3-propionic acid, β-Indolepropionic acid
InChI Formula InChI=1S/C11H11NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6H2,(H,13,14)
SMILES OC(=O)CCc1c[nH]c2ccccc12
IUPAC Name 3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 189.21
Formula Weight 189.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.