missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl acetate, 97%

Product Code. 11361767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361767 1 g 1g
11371767 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361767 Supplier Thermo Scientific Alfa Aesar Supplier No. L04932.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-08-2 | C10H9NO2 | 175.187 g/mol

Useful for detection of esterases3-Indoxyl acetate acts as a colorimetric substrate used for differentiating esterases from various microbiological species. Further, it is used as a plant growth-stimulating hormone and reagent. In addition to this, it acts as a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful for detection of esterases3-Indoxyl acetate acts as a colorimetric substrate used for differentiating esterases from various microbiological species. Further, it is used as a plant growth-stimulating hormone and reagent. In addition to this, it acts as a useful synthetic intermediate.

Solubility
Soluble in ethanol, water, chloroform, dichloromethane and methanol.

Notes
Light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-08-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00014561
InChI Key JBOPQACSHPPKEP-UHFFFAOYSA-N
Synonym 3-acetoxyindole, indoxyl acetate, 3-indoxyl acetate, indoxyl-o-acetate, 3-indolyl acetate, indole, 3-acetato, indoxylacetate, 1h-indol-3-ol, 3-acetate, 1h-indol-3-ol, acetate ester, acetic acid, 3-indolyl ester
PubChem CID 11841
IUPAC Name 1H-indol-3-yl acetate
SMILES CC(=O)OC1=CNC2=CC=CC=C21

Specifications

Melting Point 128°C to 130°C
Quantity 1 g
Beilstein 143086
Solubility Information Soluble in ethanol,water,chloroform,dichloromethane and methanol.
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 3-Indoxyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.