missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl sulfate potassium salt, 97%

Product Code. 11408956
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408956 0.25 g 0.25g
11488946 1 g 1g
11498946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408956 Supplier Thermo Scientific Alfa Aesar Supplier No. A17079.01
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2642-37-7 | C8H6KNO4S | 251.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2642-37-7
Molecular Formula C8H6KNO4S
Molecular Weight (g/mol) 251.297
MDL Number MFCD00037931
InChI Key MDAWATNFDJIBBD-UHFFFAOYSA-M
Synonym potassium 1h-indol-3-yl sulfate, urinary indican, potassium indol-3-yl sulfate, indican urinary, indol-3-yl potassium sulfate, indoxyl sulfate potassium salt, potassium indol-3-yl sulphate, unii-567hmw942w, indol-3-ol, potassium sulfate, indol-3-yl sulfate, potassium salt
PubChem CID 5177095
IUPAC Name potassium;1H-indol-3-yl sulfate
SMILES C1=CC=C2C(=C1)C(=CN2)OS(=O)(=O)[O-].[K+]

Specifications

Melting Point 165°C (decomposition)
Quantity 0.25 g
Beilstein 3921324
Sensitivity Light sensitive
Formula Weight 251.31
Percent Purity 97%
Chemical Name or Material 3-Indoxyl sulfate potassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.