missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl Sulfate Potassium Salt, TRC

Product Code. 30007170
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007170 250 mg 250mg
30012415 500 mg 500mg
30003516 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007170 Supplier Toronto Research Chemicals Supplier No. TRCI655100250MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2642-37-7
Molecular Formula C8 H6 N O4 S . K
Molecular Weight (g/mol) 251.3
Synonym 1H-Indol-3-ol, 3-(hydrogen sulfate), potassium salt (1:1) (ACI), 1H-Indol-3-ol, hydrogen sulfate (ester), monopotassium salt (9CI), Indol-3-ol, hydrogen sulfate (ester), monopotassium salt (8CI), Indol-3-yl potassium sulfate (6CI, 7CI), Indol-3-ol, potassium sulfate, Indol-3-yl sulfate, potassium salt, Indoxyl sulfate potassium salt, Potassium indol-3-yl sulfate, 3-Indoxyl sulfate potassium salt
IUPAC Name potassium;1H-indol-3-yl sulfate
SMILES [K+].[O-]S(=O)(=O)Oc1c[nH]c2ccccc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H7NO4S.K/c10-14(11,12)13-8-5-9-7-4-2-1-3-6(7)8;/h1-5,9H,(H,10,11,12);/q;+1/p-1
Formula Weight 250.9655
Chemical Name or Material 3-Indoxyl Sulfate Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.