missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1-methyl-1H-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10713264
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10713264 1 g Amber Glass Bottle 1g
10652665 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713264 Supplier Thermo Scientific Maybridge Supplier No. CC48311DA
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 92525-10-5
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.002
MDL Number MFCD08435890
InChI Key SITJXRWLFQGWCB-UHFFFAOYSA-N
PubChem CID 13334057
IUPAC Name 3-iodo-1-methylpyrazole
SMILES CN1C=CC(=N1)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-iodo-1-methyl-1h-pyrazole, 1-methyl-3-iodopyrazole, 1h-pyrazole, 3-iodo-1-methyl, 3-iodo-1-methy-1h-pyrazole, iodo-n-methylpyrazole, 3-idio-1-methylpyrazole, 3-iodanyl-1-methyl-pyrazole, 1h-pyrazole,3-iodo-1-methyl, 3 5-iodopyrazole, 1-me
Formula Weight 208
Percent Purity 97%
Chemical Name or Material 3-Iodo-1-methyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.