missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 15472729
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472729 1 g 1g
15482729 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472729 Supplier Thermo Scientific Alfa Aesar Supplier No. H66356.03
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92525-10-5 | Molecular Formula: C4H5IN2 | Molecular Weight: 208.002 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92525-10-5
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.002
MDL Number MFCD08435890
InChI Key SITJXRWLFQGWCB-UHFFFAOYSA-N
Synonym 3-iodo-1-methyl-1h-pyrazole, 1-methyl-3-iodopyrazole, 1h-pyrazole, 3-iodo-1-methyl, 3-iodo-1-methy-1h-pyrazole, iodo-n-methylpyrazole, 3-idio-1-methylpyrazole, 3-iodanyl-1-methyl-pyrazole, 1h-pyrazole,3-iodo-1-methyl, 3 5-iodopyrazole, 1-me
PubChem CID 13334057
IUPAC Name 3-iodo-1-methylpyrazole
SMILES CN1C=CC(=N1)I

Specifications

Color Brown
Density 1.936 g/mL
Flash Point 110°C (230°F)
Refractive Index 1.588
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 208
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Iodo-1-methyl-1H-pyrazole

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.