missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1H-pyrazole, 97%

Product Code. 15477657
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15477657 1 g 1g
15487657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477657 Supplier Thermo Scientific Alfa Aesar Supplier No. H36465.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4522-35-4 | C3H3IN2 | 193.98 g/mol

3-Iodo-1H-pyrazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-1H-pyrazole is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4522-35-4
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.98
MDL Number MFCD12022324,MFCD01463487
InChI Key RUKDVLFJSMVBLV-UHFFFAOYSA-N
Synonym 3-iodo-1h-pyrazole, 3-iodopyrazole, iodopyrazole, 1h-pyrazole, 3-iodo, 3-iodo-2h-pyrazole, iodo-pyrazole, 5-iodopyrazole, acmc-2097sa
PubChem CID 1239830
IUPAC Name 5-iodo-1H-pyrazole
SMILES IC1=CC=NN1

Specifications

Melting Point 69°C to 72°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 193.98
Percent Purity 97%
Chemical Name or Material 3-Iodo-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.