missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 11966231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11966231 5 g 5g
11956231 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966231 Supplier Thermo Scientific Acros Supplier No. 444810050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4522-35-4 | Molecular Formula: C3H3IN2 | Molecular Weight: 193.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4522-35-4
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.98
MDL Number MFCD12022324,MFCD01463487
InChI Key RUKDVLFJSMVBLV-UHFFFAOYSA-N
Synonym 3-iodo-1h-pyrazole, 3-iodopyrazole, iodopyrazole, 1h-pyrazole, 3-iodo, 3-iodo-2h-pyrazole, iodo-pyrazole, 5-iodopyrazole, acmc-2097sa
PubChem CID 1239830
IUPAC Name 5-iodo-1H-pyrazole
SMILES IC1=CC=NN1

Specifications

Melting Point 68.0°C to 73.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 193.98
Percent Purity 97%
Chemical Name or Material 3-Iodo-1H-pyrazole
compliance-icons
Product Identifier
  • 3-Iodo-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.