missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Iodo-L-tyrosine, 97%

Product Code. 10289533 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10289533 25 g Glass Bottle 25g
10285160 1 g Glass Bottle 1g
10279533 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289533 Supplier Thermo Scientific Chemicals Supplier No. 122440250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-78-0 | C9H10INO3 | 307.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-78-0
Molecular Formula C9H10INO3
Molecular Weight (g/mol) 307.09
MDL Number MFCD00002608
InChI Key UQTZMGFTRHFAAM-JLDDOWRYNA-N
Synonym 3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
PubChem CID 439744
ChEBI CHEBI:27847
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 196.0°C to 198.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Specific Rotation − 3.18
Merck Index 15, 5090
Solubility Information Solubility in water: .. Other solubilities: soluble in 15 parts boiling water
Water 1% max. (K.F.)
Quantity 25 g
Formula Weight 307.08
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Iodo-L-tyrosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.