missing translation for 'onlineSavingsMsg'
Learn More

3-Iodochromone, 97%

Product Code. 15497227
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497227 1 g 1g
15407237 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497227 Supplier Thermo Scientific Alfa Aesar Supplier No. H34295.03
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122775-34-2 | C9H5IO2 | 272.04 g/mol

3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodochromone is used in the synthesis of 2-(N-azolyl)chromones, 3-azolylthiochromones.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agent, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122775-34-2
Molecular Formula C9H5IO2
Molecular Weight (g/mol) 272.04
MDL Number MFCD09475514
InChI Key SZDBDDKQJHHRGH-UHFFFAOYSA-N
Synonym 3-iodochromone, 3-iodo-4h-chromen-4-one, 3-iodo-4h-benzopyran-4-one
PubChem CID 10901720
IUPAC Name 3-iodo-4H-chromen-4-one
SMILES IC1=COC2=CC=CC=C2C1=O

Specifications

Melting Point 95°C to 97°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 272.04
Percent Purity 97%
Chemical Name or Material 3-Iodochromone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.