missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopropionic acid, 99%

Product Code. 11327785
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327785 5 g 5g
11337785 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327785 Supplier Thermo Scientific Alfa Aesar Supplier No. L02655.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-76-4 | C3H5IO2 | 199.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-76-4
Molecular Formula C3H5IO2
Molecular Weight (g/mol) 199.98
MDL Number MFCD00002765
InChI Key KMRNTNDWADEIIX-UHFFFAOYSA-N
Synonym 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid
PubChem CID 8856
IUPAC Name 3-iodopropanoic acid
SMILES OC(=O)CCI

Specifications

Melting Point 80°C to 83°C
Quantity 5 g
UN Number UN3261
Beilstein 1699514
Sensitivity Light sensitive
Formula Weight 199.98
Percent Purity 99%
Chemical Name or Material 3-Iodopropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.