missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopyridine, 99%

Product Code. 11450837
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450837 25 g 25g
11440837 5 g 5g
11460837 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450837 Supplier Thermo Scientific Alfa Aesar Supplier No. B23225.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1120-90-7 | C5H4IN | 204.998 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-90-7
Molecular Formula C5H4IN
Molecular Weight (g/mol) 204.998
MDL Number MFCD00023553
InChI Key XDELKSRGBLWMBA-UHFFFAOYSA-N
Synonym 3-iodo pyridine, 3-iodo-pyridine, pyridine, 3-iodo, 3-iodpyridin, 3-iodopyridine, pubchem6634, acmc-1bvap, ksc490s0t
PubChem CID 70714
IUPAC Name 3-iodopyridine
SMILES C1=CC(=CN=C1)I

Specifications

Melting Point 53°C to 56°C
Boiling Point 90°C to 92°C (14 mmHg)
Flash Point 106°C (222°F)
Quantity 25 g
Beilstein 106059
Sensitivity Light sensitive
Formula Weight 205
Percent Purity 99%
Chemical Name or Material 3-Iodopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.