missing translation for 'onlineSavingsMsg'
Learn More

3-iodotetrahydrofuran, 97%, Thermo Scientific™

Product Code. 10285474
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10285474 1 g Amber Glass Bottle 1g
10623455 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285474 Supplier Thermo Scientific Maybridge Supplier No. CC76011DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 121138-01-0
Molecular Formula C4H7IO
Molecular Weight (g/mol) 198.00
MDL Number MFCD09878873
InChI Key BKIQORJIKOPRCG-UHFFFAOYNA-N
Synonym 3-iodotetrahydrofuran, 3-iodo-tetrahydro-furan, furan, tetrahydro-3-iodo, 3-iodanyloxolane, iodotetrahydrofuran
PubChem CID 22139264
IUPAC Name 3-iodooxolane
SMILES IC1CCOC1

Specifications

CAS Min % 96.5
Color Colorless
CAS Max % 100.0
Quantity 1 g
Packaging Amber Glass Bottle
Physical Form Liquid
Assay Percent Range 97%
Boiling Point 110°C
Formula Weight 198
Percent Purity 97%
Chemical Name or Material 3-Iodotetrahydrofuran
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.