missing translation for 'onlineSavingsMsg'
Learn More

3-Isobutyl-1-methyl-1H-pyrazole-5-carboxylic Acid, TRC

Product Code. 30329431
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329431 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329431 Supplier Toronto Research Chemicals Supplier No. TRCB448783500MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Isobutyl-1-methyl-1H-pyrazole-5-carboxylic Acid
CAS 769132-77-6
Molecular Formula C9 H14 N2 O2
Quantity 500 mg
InChI Formula InChI=1S/C9H14N2O2/c1-6(2)4-7-5-8(9(12)13)11(3)10-7/h5-6H,4H2,1-3H3,(H,12,13)
SMILES CC(C)Cc1cc(C(=O)O)n(C)n1
IUPAC Name 2-methyl-5-(2-methylpropyl)pyrazole-3-carboxylic acid
Molecular Weight (g/mol) 182.22
Formula Weight 182.106
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.