missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isobutyl-1-Methylxanthine, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
Description
A non-specific inhibitor of adenosine 3',5'-cyclic monophosphate phosphodiesterase (cAMP PDE)4, significantly more effective than theophylline. Also inhibits cGMP phosphodiesterases. IBMX inhibits cyclic nucleotide PDE with subsequent inhibition of cyclic nucleotide hydrolysis, resulting in accumulation of cyclic AMP and guanosine 3',5'-cyclic monophosphate.
- Inhibits phenylephrine-induced release of 5-hydroxytryptamine from neuroendocrine epithelial cells of the airway mucosa (IC50: 1.3μM)
- Serves as an adenosine receptor antagonist
- Inhibits ion channels in the neuromuscular junction, GH3 cells, and vascular smooth muscle cells
- Induces calcium release from intracellular stores in sensory neurons
- Inhibits the growth of carcinoma cells both in vivo and in vitro in mice
Specifications
Specifications
| Melting Point | 195°C to 202°C |
| Quantity | 1 g |
| Formula Weight | 222.25 |
| Physical Form | Powder |
Safety and Handling
Product Identifier
- 3-Isobutyl-1-methylxanthine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 3-Isobutyl-1-methylxanthine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction