missing translation for 'onlineSavingsMsg'
Learn More

3-Isobutyl-1-Methylxanthine, MP Biomedicals™

Product Code. 11485222 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485222 100 mg 100mg
11495222 250 mg 250mg
11405232 1 g 1g
11415232 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11485222 Supplier MP Biomedicals Supplier No. 02195262.1
  • ONLINE EXCLUSIVE PRICE
    56.50 €
    53.55 € / 100mg
    Save 2.95 €
    5% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Known as IBMX. 3-Isobutyl-1-Methylxanthine, MP Biomedicals is a potent, non-specific inhibitor of cAMP and cGMP phosphodiesterases, significantly more effective than theophylline. This inhibition activates PKA, leading to decreased proliferation, increase

A non-specific inhibitor of adenosine 3',5'-cyclic monophosphate phosphodiesterase (cAMP PDE)4, significantly more effective than theophylline. Also inhibits cGMP phosphodiesterases. IBMX inhibits cyclic nucleotide PDE with subsequent inhibition of cyclic nucleotide hydrolysis, resulting in accumulation of cyclic AMP and guanosine 3',5'-cyclic monophosphate.

  • Inhibits phenylephrine-induced release of 5-hydroxytryptamine from neuroendocrine epithelial cells of the airway mucosa (IC50: 1.3μM)
  • Serves as an adenosine receptor antagonist
  • Inhibits ion channels in the neuromuscular junction, GH3 cells, and vascular smooth muscle cells
  • Induces calcium release from intracellular stores in sensory neurons
  • Inhibits the growth of carcinoma cells both in vivo and in vitro in mice
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28822-58-4
Molecular Formula C10H14N4O2
Molecular Weight (g/mol) 222.25
MDL Number MFCD00005584
InChI Key APIXJSLKIYYUKG-UHFFFAOYSA-N
Synonym 3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine
PubChem CID 3758
ChEBI CHEBI:43253
IUPAC Name 1-methyl-3-(2-methylpropyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O

Specifications

Melting Point 195°C to 202°C
Quantity 100 mg
Formula Weight 222.25
Physical Form Powder
compliance-icons
Product Identifier
  • 3-Isobutyl-1-methylxanthine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3-Isobutyl-1-methylxanthine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.