missing translation for 'onlineSavingsMsg'
Learn More

3-Isocyanato-5-methyl-2-phenylfuran, 97%, Thermo Scientific™

Product Code. 10678694
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10678694 1 g Amber Glass Bottle 1g
10002124 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10678694 Supplier Thermo Scientific Maybridge Supplier No. CC49706DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-18-4
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD09025851
InChI Key GDLFJANXKGIZKA-UHFFFAOYSA-N
Synonym 5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
PubChem CID 18525836
IUPAC Name 3-isocyanato-5-methyl-2-phenylfuran
SMILES CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O

Especificaciones

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 3-Isocyanato-5-methyl-2-phenylfuran
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.