missing translation for 'onlineSavingsMsg'
Learn More

3-Isoniazid, TRC

Product Code. 30137804
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137804 10 g 10g
17901345 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137804 Supplier Toronto Research Chemicals Supplier No. TRCI82146010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-53-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
Synonym Nicotinic Acid Hydrazide, Nicotinohydrazide, 3-Pyridinecarboxylic Acid Hydrazide, (3-Pyridylcarbonyl)hydrazine, 3-Pyridinecarbohydrazide, 3-Pyridinecarboxylic Hydrazide, 3-Pyridoylhydrazine;
IUPAC Name pyridine-3-carbohydrazide
SMILES C(C=1C=CC=NC1)(=O)NN.Cl.Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H7N3O/c7-9-6(10)5-2-1-3-8-4-5/h1-4H,7H2,(H,9,10)
Formula Weight 137.0589
Chemical Name or Material 3-Isoniazid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.