missing translation for 'onlineSavingsMsg'
Learn More

3-Ketopalmitoyl-CoA, TRC

Product Code. 30395890
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.50mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395890 0.5 mg 0.50mg
30395760 1 mg 1mg
30406225 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395890 Supplier Toronto Research Chemicals Supplier No. TRCK1980030.5MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34619-89-1
Molecular Formula C37H64N7O18P3S
Molecular Weight (g/mol) 1019.93
Synonym 3-Oxohexadecanoyl-CoA, 3-Oxopalmitoyl-CoA, S-(3-Ketohexadecanoyl)coenzyme A, beta-Ketopalmitoyl-coenzyme A
IUPAC Name S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 3-oxohexadecanethioate
SMILES CCCCCCCCCCCCCC(=O)CC(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP(=O)(O)OP(=O)(O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C37H64N7O18P3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-25(45)20-28(47)66-19-18-39-27(46)16-17-40-35(50)32(49)37(2,3)22-59-65(56,57)62-64(54,55)58-21-26-31(61-63(51,52)53)30(48)36(60-26)44-24-43-29-33(38)41-23-42-34(29)44/h23-24,26,30-32,36,48-49H,4-22H2,1-3H3,(H,39,46)(H,40,50)(H,54,55)(H,56,57)(H2,38,41,42)(H2,51,52,53)/t26-,30-,31-,32+,36-/m1/s1
Formula Weight 1019.3241
Chemical Name or Material 3-Ketopalmitoyl-CoA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.