missing translation for 'onlineSavingsMsg'
Learn More

3-Maleimidopropionic acid, 95%

Product Code. 15438098
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438098 5 g 5g
15428098 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438098 Supplier Thermo Scientific Alfa Aesar Supplier No. H61536.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7423-55-4 | C7H7NO4 | 169.14 g/mol

3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

Solubility
Soluble in methanol, dimethyl formamide, alcohol and dimethyl sulfoxide. Slightly soluble in water.

Notes
Moisture sensitive. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-55-4
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00043030
InChI Key IUTPJBLLJJNPAJ-UHFFFAOYSA-N
Synonym 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw
PubChem CID 573621
IUPAC Name 3-(2,5-dioxopyrrol-1-yl)propanoic acid
SMILES OC(=O)CCN1C(=O)C=CC1=O

Specifications

Melting Point 108°C to 112°C
Color White
Quantity 5 g
Beilstein 1528952
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol,dimethyl formamide,alcohol and dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 169.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Maleimidopropionic acid
compliance-icons
Product Identifier
  • 3-Maleimidopropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Maleimidopropionicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.